Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ameloblastin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113434
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113434 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113434 Supplier Invitrogen™ Supplier No. PA5113434
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ameloblastin Polyclonal Antibody for Western Blot, ELISA

This gene encodes the nonamelogenin enamel matrix protein ameloblastin. The encoded protein may be important in enamel matrix formation and mineralization. This gene is located in the calcium-binding phosphoprotein gene cluster on chromosome 4. Mutations in this gene may be associated with dentinogenesis imperfect and autosomal dominant amylogenesis imperfect.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ameloblastin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene AMBN
Gene Accession No. Q62840, Q9NP70
Gene Alias AI1F; AMBN; Amelin; ameloblastin; ameloblastin (enamel matrix protein); ameloblastin precursor; enamel matrix protein
Gene Symbols AMBN
Host Species Rabbit
Immunogen Recombinant Human Ameloblastin protein (264-387AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25376, 258
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.