Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ameloblastin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103108
Description
Antibody detects endogenous levels of total Ameloblastin.
This gene encodes the nonamelogenin enamel matrix protein ameloblastin. The encoded protein may be important in enamel matrix formation and mineralization. This gene is located in the calcium-binding phosphoprotein gene cluster on chromosome 4. Mutations in this gene may be associated with dentinogenesis imperfect and autosomal dominant amylogenesis imperfect.
Specifications
Ameloblastin | |
Polyclonal | |
Unconjugated | |
AMBN | |
AI1F; AMBN; Amelin; ameloblastin; ameloblastin (enamel matrix protein); ameloblastin precursor; enamel matrix protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11698, 25376, 258 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O55189, Q62840, Q9NP70 | |
AMBN | |
A synthesized peptide derived from human AMBN(Accession Q9NP70), corresponding to amino acid residues A266-P316. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction