Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AMELX Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | AMELX |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5118945
|
Invitrogen™
PA5118945 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5122244
|
Invitrogen™
PA5122244 |
20 μL |
Each for $209.50
|
|
|||||
Description
AMELX Polyclonal Antibody for Western Blot
This gene encodes a member of the amelogenin family of extracellular matrix proteins. Amelogenins are involved in biomineralization during tooth enamel development. Mutations in this gene cause X-linked amelogenesis imperfecta. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
AMELX | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
AMELX | |
AI1E; AIH1; ALGN; Amel; amelogenesis imperfecta 1; amelogenin (amelogenesis imperfecta 1, X-linked); amelogenin (X chromosome, amelogenesis imperfecta 1); amelogenin X chromosome; amelogenin, X isoform; amelogenin, X-linked; Amelx; AMG; AMGL; AMGX; enamel matrix protein; leucine-rich amelogenin peptide; LRAP; M100888; Rgsc888 | |
AMELX | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q99217 | |
265 | |
Recombinant protein Amelogenin, X-Linked. The antigen corresponds to amino acid range 49-187 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title