Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMER1 Monoclonal Antibody (729419)
GREENER_CHOICE

Catalog No. PIMA524343
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524343 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524343 Supplier Invitrogen™ Supplier No. MA524343
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

The AMER1 gene encodes the protein APC membrane recruitment 1, also known as Wilms tumor 1-associating protein (WT1-associating protein). This protein plays a significant role in the Wnt signaling pathway, which is critical for cellular processes such as proliferation, differentiation, and migration. AMER1 interacts with the adenomatous polyposis coli (APC) protein and is involved in the regulation of β-catenin, a key mediator of Wnt signaling. By recruiting APC to the cell membrane, AMER1 facilitates the degradation of β-catenin, thereby acting as a negative regulator of the pathway. This function is vital in maintaining cellular homeostasis and preventing uncontrolled cell growth. Mutations or dysregulation of AMER1 are associated with various cancers, including colorectal cancer, as aberrant Wnt signaling can lead to oncogenesis. Moreover, germline mutations in AMER1 are linked to conditions such as osteoporosis-pseudoglioma syndrome, highlighting its role in bone development and eye health. Ongoing research focuses on elucidating the detailed mechanisms of AMER1's involvement in signaling pathways and its potential as a therapeutic target in cancer and genetic disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMER1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 729419
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene AMER1
Gene Accession No. Q5JTC6
Gene Alias 2810002O09Rik; adenomatous polyposis coli membrane recruitment 1; Amer1; APC membrane recruitment 1; APC membrane recruitment protein 1; AW492303; Fam123b; family with sequence similarity 123, member B; family with sequence similarity 123B; OSCS; protein Fam123b; RGD1560322; RP11-403E24.2; Wilms tumor gene on the X chromosome protein; Wilms tumor on the X; WTX
Gene Symbols AMER1
Host Species Mouse
Immunogen E. coli-derived recombinant human WTX Met421-Leu573.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139285
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.