Learn More
Invitrogen™ AMER1 Polyclonal Antibody

Description
AMER1 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | AMER1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | AMER1 |
| Gene Accession No. | Q5JTC6, Q7TS75 |
| Gene Alias | 2810002O09Rik; adenomatous polyposis coli membrane recruitment 1; Amer1; APC membrane recruitment 1; APC membrane recruitment protein 1; AW492303; Fam123b; family with sequence similarity 123, member B; family with sequence similarity 123B; OSCS; protein Fam123b; RGD1560322; RP11-403E24.2; Wilms tumor gene on the X chromosome protein; Wilms tumor on the X; WTX |
| Gene Symbols | AMER1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.