Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMER2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571433
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571433 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571433 Supplier Invitrogen™ Supplier No. PA571433
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AMER2 Polyclonal Antibody for Western Blot

Negative regulator of the canonical Wnt signaling pathway involved in neuroectodermal patterning. Acts by specifically binding phosphatidylinositol 4,5-bisphosphate (PtdIns(4,5)P2), translocating to the cell membrane and interacting with key regulators of the canonical Wnt signaling pathway, such as components of the beta-catenin destruction complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMER2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene AMER2
Gene Accession No. Q8N7J2
Gene Alias 2600011E07Rik; AMER2; APC membrane recruitment 2; APC membrane recruitment protein 2; Fam123a; family with sequence similarity 123, member A; family with sequence similarity 123A; protein FAM123A; RGD1559605
Gene Symbols AMER2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human AMER2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219287
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.