Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amerex™ High-Performance Fire Extinguishers: Fast Flow
Specifications
Product Type | Fire Extinguisher |
---|---|
Material | Steel |
For Use With (Application) | Industrial fires |
Includes | Wall bracket |
Range | 40 to 50 ft. |
Catalog Number | Mfr. No. | Discharge Time | Weight (English) | Reagent Type | UL Rating | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Discharge Time | Weight (English) | Reagent Type | UL Rating | Price | Quantity & Availability | ||
19-860-511
|
Amerex™
9592 |
18 sec. | 20 lb. | ABC dry chemical | 4A:40B:C | N/A | N/A | N/A | |
Description
- 240 psi operating pressure
- Painted, galvanized drawn-steel cylinder with gauge guards and all-metal valve construction
- Barcoded and bilingual labels
- Provide high-flow rate, fast discharge for quicker knockdown and operator protection
- Range: 12.2 to 15.2m (40 to 50 ft.)
- Temperature range: -54° to +49°C (-65° to +120°F)
Compliance
NFPA 10 Standard.
Warranty and Services
12 years
Specifications
Fire Extinguisher | |
Industrial fires | |
40 to 50 ft. | |
-53.8°C to +48.8°C |
Steel | |
Wall bracket | |
-65° to +120°F |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title