Learn More
American Educational Products Buret
SCI_ED
Catalog No. S04319
Click to view available options
:
w/PTFE Stopcock; Freeze-proof stopcock; Capacity: 50mL
This item is not returnable. View return policy
This item is not returnable. View return policy
High quality borosilicate glass burette with PTFE Stopcock is perfect of students.
  • 50mL Capacity
  • Freeze-proof stopcock
Specifications
Product Type Buret
Includes Non-stick ptfe stopcock
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.