Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Educational Products Compound Bar
SCI_ED

Catalog No. S41704
Click to view available options
:
Compound Bar

Includes

Instructions

Made from different metals to simulate a typical thermostat.

  • Bar is bimetallic and has an attached handle
  • When heated, the bar bends dramatically

Order Info

Dimensions: 10 x 0.75 x 0.75 in. (25 x 1.9 x 1.9cm); weight: 0.2 lb. (90g).

Specifications

Product Type Compound Bar
For Use With (Application) To demonstrate the thermostat principle
Includes Instruction
Weight (English) 0.2 lb.
Weight (Metric) 90 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.