Learn More
American Safety Razors™ American Line Extra Keen 3 Facet, Single Edge Razor Blades, Aluminum Back
Click to view available options
Quantity:
5000 Blades
Description
Resealable clear storage box with full access hinged lid
- Blue 0.009 in. high carbon steel
- Special triple-facet edge for critical cuts
- Each blade protected in shelling paper
- Non-rust aluminum back
- Razor fits all standard razor blade tools
- Double hang tags holds 100 single edge blades
Specifications
Specifications
Blade Material | Carbon Steel |
Product Type | Razor Blade |
Quantity | 5000 Blades |