Learn More
American Scientific Lenz Law Apparatus
SDPSCI_ED
Catalog No. s12862
Click to view available options
:
Lenz Law Apparatus
This item is not returnable. View return policy
This item is not returnable. View return policy
To demonstrate electric and magnetic forces.
Introduction: Two identical looking plugs are dropped through the copper pipe but react very differently. As you watch them drop through the viewing window you will see one drop quickly and without resistance while the other moves slowly through the tube.
Specifications
Product Type Lenz Law Apparatus
Includes Copper tube, magnet plug, metal plug, end caps, and a Teacher's Guide.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.