Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Scientific Solid Steel Ball (Smashing Steel Sphere Kinetic Energy Demo)
SDPSCI_ED

Catalog No. S12857
Click to view available options
:
Steel solid ball (Pair)
This item is not returnable. View return policy
This item is not returnable. View return policy

For physics experiments and demonstrations such as kinetic, density and volume measurements, pendulum bob, potential energy and conservation experiments, ballistics.

  • Diameter: 40mm (1.5 in.)
  • Sold as pair
  • Includes teacher's guide

Specifications

Product Type Solid Ball (Pair)
Includes Teacher's guide
Material Steel
Size Dia: 1.5 in. (40mm)
Diameter (English) Outer 1.5 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.