Learn More
Amersham™ CyDye™ 800 Goat-Anti-Mouse Secondary Antibody, Cytiva
Amersham™ CyDye™ NIR-labeled secondary antibodies are designed for quantitative fluorescent Western blotting.
$190.08 - $323.95
Specifications
Applications | Western Blot |
---|---|
Conjugate | CyDye 800 |
Primary or Secondary | Secondary |
Content And Storage | 2°C to 8°C, protected from light |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
09-928-133
|
Cytiva
29360788 |
0.1 mg |
Each for $190.08
|
|
|||||
09-928-134
|
Cytiva
29360789 |
0.5 mg |
Each for $323.95
|
|
|||||
Description
Fluorescence detection is a direct Western blotting method in which a secondary antibody is conjugated to a fluorophore, avoiding the need for ancillary detection reagents. Different fluorophores are available for signal detection at either visible light or near-infrared (NIR) wavelengths.
Amersham CyDye 700 and 800 secondary antibodies are labeled with NIR fluorophores that emit light at wavelengths of 700 nm or 800 nm. By using Amersham CyDye 700 and 800 secondary antibodies together with a pair of appropriate rabbit or mouse primary antibodies, you can perform multiplexed experiments.
Key benefits:
- Easy handling and a strong signal-to-noise ratio yield reproducible results
- Multiplexing analysis of more than one target protein on a single blot increases process efficiency
- Total protein normalization (TPN) generates reliable and accurate quantitation
- Stable signals allow membranes to be stored and rescanned for future studies
Specifications
Western Blot | |
CyDye 800 | |
Secondary | |
2°C to 8°C, protected from light |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.