Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Amersham™ CyDye™ Antibody Labeling Kits
Click to view available options
Label or Dye:
Cy3 Reactive Dye
Cy5 Reactive Dye
Product Type:
Cy3 Ab Labeling Kit
Cy5 Ab Labeling Kit
Description
CyDye fluorescent dyes are supplied as highly water soluble, reactive NHS esters for easy conjugation to biomolecules containing aliphatic amino groups.
- Conjugations are carried out under mild conditions and are complete within 1 hour
- Dyes are packaged in premeasured amounts and foil sealed to ensure consistent labelings
Order Info
Antibody kits contain all reagents required for 2 x 1mg reactions. Monoclonal antibody kits contain all reagents required for 2 x 0.1mg reactions.

Specifications
Specifications
For Use With (Application) | For the conjugation of proteins with Cy5 Reactive Dye |
Quantity | Each |
Product Type | Cy5 Ab Labeling Kit |
pH Range | 7 to 7.3 |
Molecular Weight (g/mol) | 975.15 |
Solubility | Cy5 dye: Soluble in cold water and hot water Coupling buffer: Soluble in cold water and hot water Elution buffer: Soluble in cold water and hot water |
Content And Storage | 2°C to 8°C, protected from light |
Label or Dye | Cy5 Reactive Dye |
Color | Blue |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction