Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham CyDye Fluorescent Nucleotides
GSA_VA

Catalog No. 45200578 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
250 nmol
250 nmol (5 x 25 nmol Cy3-dCTP + 5 x 25 nmol Cy5-dCTP)
Label or Dye:
Orange
Orange/Far-red
Catalog No. Quantity Label or Dye
45-200-578 250 nmol (5 x 25 nmol Cy3-dCTP + 5 x 25 nmol Cy5-dCTP) Orange/Far-red
45-200-576 250 nmol Orange
2 options

Catalog No. 45-200-578

Supplier: Cytiva PA55321

Ideal for multiplexing within a single sample. Cytiva Amersham CyDye Fluorescent Nucleotides are fluorescent dyes with superior characteristics for genomic and protein research. Available in multiple sizes and colors, including a multipack option for maximum versatility.

CyDyes are a group of bright fluorescent dyes which can be used in genomic and protein research. They are ideal for labeling DNA and RNA probes, especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping.CyDye labeled nucleotides are available in a range of bright, intense colors with narrow emission bands, well suited for multiplexing within a single sample. They exhibit the following properties:
  • High sensitivity
  • Low nonspecific binding
  • High photostability
  • High water solubility
  • pH insensitive

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 250 nmol (5 x 25 nmol Cy3-dCTP + 5 x 25 nmol Cy5-dCTP)
Label or Dye Orange/Far-red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.