Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ CyDye™ Fluorescent Nucleotides

Catalog No. 45001238 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
Cy3-CTP
Cy3-UTP
Cy3-dCTP
Cy3-dUTP
Cy3-dUTP, Cy5-dUTP
Cy3.5-dCTP
Cy5-CTP
Cy5-UTP
Cy5-dCTP
Cy5-dUTP
Quantity:
100 nm
100 nmol
25 nmol
5 x 25 nmol
Color:
Far-Red
Orange
Orange, Far-Red
Scarlet
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity Color
45-001-238 Cy3-UTP 100 nmol Orange
45-000-738 Cy3-dUTP 25 nmol Orange
45-000-737 Cy3-dCTP 25 nmol Orange
45-000-739 Cy5-dCTP 25 nmol Far-Red
45-000-740 Cy5-dUTP 25 nmol Far-Red
45-500-149 Cy3.5-dCTP 25 nmol Scarlet
45-001-290 Cy3-CTP 100 nm Orange
45-001-291 Cy5-CTP 100 nmol Far-Red
45-001-239 Cy5-UTP 100 nmol Far-Red
45-001-241 Cy3-dUTP, Cy5-dUTP 5 x 25 nmol Orange, Far-Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 45-001-238 Supplier Cytiva Supplier No. PA53026

Label DNA and RNA probes. Cytiva Life Sciences™ Amersham™ CyDye™ Fluorescent Nucleotides are optimized for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping.

For labeling DNA and RNA probes especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping. CyDye labeled nucleotides are available in a range of bright, intense colors with narrow emission bands—ideal for multiplexing within a single sample.

  • Low nonspecific binding
  • High photostability
  • High water solubility
  • pH insensitive

TRUSTED_SUSTAINABILITY

Specifications

Description CyDyes are a group of bright fluorescent dyes which can be used in genomic and protein research. Cy dye labelled nucleotides exhibit the following properties:
pH 7
Molecular Weight (g/mol) 765.95 (Monofunction), 949.11 (Bisfunction)
Product Type Cy3-UTP
Reagents Fluorescent Labeling Reagents
Wavelength 570nm
Content And Storage -15°C to -30°C
Quantity 100 nmol
Color Orange
For Use With (Application) Genomic and Protein research, labeling DNA and RNA probes, especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping and gene mapping
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.