Learn More
Description
The use of mono-reactive maleimides is particularly suited to antibody labeling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired.
- Suitable for the labeling of antibodies
- Example protocols included with each dye pack
Antibody labeling
Specifications
Specifications
| For Use With (Application) | For labelling via thiol groups For use in steady state fluorescence, FP and FRET applications |
| Quantity | 1 mg |
| Product Type | Cy5.5 Maleimide |
| Kit Contents | Foil packs, each containing 1, 5 or 25mg dried dye |
| pH Range | 3 to 10 |
| Molecular Weight (g/mol) | 1128.42 |
| Solubility | Soluble in cold water and hot water |
| Content And Storage | 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles. |
| Label or Dye | Cy5.5 |
| Color | Dark Blue |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.