Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive Maleimide)
GSA_VA

Catalog No. 45500144 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Label or Dye:
Cy3
Cy5
Cy5.5
Product Type:
Cy3 Maleimide
Cy5 Maleimide
Cy5.5 Maleimide
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Label or Dye Product Type
45-500-144 25 mg Cy5 Cy5 Maleimide
45-001-255 5 mg Cy5 Cy5 Maleimide
45-001-202 1 mg Cy3 Cy3 Maleimide
45-001-203 1 mg Cy5 Cy5 Maleimide
09-928-087 1 mg Cy5.5 Cy5.5 Maleimide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 45-500-144 Supplier Cytiva Supplier No. PA15136

For the selective labeling of free sulfhydryl groups such as cysteine residues in peptides, proteins, and oligonucleotides

The use of mono-reactive maleimides is particularly suited to antibody labeling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired.

  • Suitable for the labeling of antibodies
  • Example protocols included with each dye pack

Applications:

Antibody labeling

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For labelling via thiol groups For use in steady state fluorescence, FP and FRET applications
Quantity 25 mg
Product Type Cy5 Maleimide
Kit Contents Foil packs, each containing 1, 5 or 25mg dried dye
pH Range 3 to 10
Molecular Weight (g/mol) 791.95
Solubility Soluble in cold water and hot water
Content And Storage 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
Label or Dye Cy5
Color Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.