Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive Maleimide)
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Label or Dye:
Cy3
Cy5
Cy5.5
Product Type:
Cy3 Maleimide
Cy5 Maleimide
Cy5.5 Maleimide
Description
The use of mono-reactive maleimides is particularly suited to antibody labeling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired.
- Suitable for the labeling of antibodies
- Example protocols included with each dye pack
Antibody labeling

Specifications
Specifications
For Use With (Application) | For labelling via thiol groups For use in steady state fluorescence, FP and FRET applications |
Quantity | 1 mg |
Product Type | Cy5.5 Maleimide |
Kit Contents | Foil packs, each containing 1, 5 or 25mg dried dye |
pH Range | 3 to 10 |
Molecular Weight (g/mol) | 1128.42 |
Solubility | Soluble in cold water and hot water |
Content And Storage | 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles. |
Label or Dye | Cy5.5 |
Color | Dark Blue |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction