Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive NHS Ester)

Catalog No. 45001263 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Label or Dye:
Cy5
Cy7
Product Type:
Cy5 NHS Ester
Cy7 NHS Ester
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Label or Dye Product Type
45-001-263 1 mg Cy7 Cy7 NHS Ester
45-000-705 50 mg Cy5 Cy5 NHS Ester
45-000-706 10 mg Cy5 Cy5 NHS Ester
45-001-267 10 mg Cy7 Cy7 NHS Ester
45-000-704 5 mg Cy5 Cy5 NHS Ester
45-001-190 1 mg Cy5 Cy5 NHS Ester
45-500-143 25 mg Cy5 Cy5 NHS Ester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 45-001-263 Supplier Cytiva Supplier No. PA17101
Add to Cart
Add to Cart

Use for highly accurate labeling of amine groups. Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive NHS Ester) contain derivitized CyDyes that carry only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies, peptides, or oligonucleotides.

  • Contains derivitized CyDyes that carry only one reactive group on each dye molecule
  • pH range: 3 to 10
  • DMSO tolerant
  • Photostable
  • Example protocols included with each dye pack

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For labeling via Amine groups, CyDyes are suitable for use in applications such as Fluorescence resonance energy transfer (FRET), time resolved-Fluorescence resonance energy transfer (TR-FRET) and Fluorescence polarization (FP)
Quantity 1 mg
Product Type Cy7 NHS Ester
Kit Contents Foil packs, each containing 1, 5, 10, 25 or 50mg dried dye
pH Range 3 to 10
Content And Storage 2°C to 8°C
Label or Dye Cy7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.