Learn More
Description
Includes
45-000-922 and 45-000-923: Labeling reagent; Crosslinker; Control DNA; Blocking agent; ECL detection reagents and ECL Gold hybridization buffer
- Direct probe labeling in a 10 min reaction, 1 h from hybridization to detection with ECL Direct, Hybond N+, and Hyperfilm ECL.
- Eliminates handling, waste, and regulatory issues associated with the use of radioactivity.
- No need to strip blots before reprobing.
- For fast and easy detection of medium- to high-target amounts in applications such as colony/plaque screens, dot blots, and PCR product analyses.
- Consistent results combining strong signals with very low backgrounds.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Colony or plaque screens, dot blots and PCR product analyses |
| Includes | DNA labeling reagent 0.5mL; Glutaraldehyde solution 0.5mL; Water 1.0mL; l Hind III control DNA (10μg/mL) 0.1mL; Gold hybridization buffer 500mL; Blocking agent 25g; ECL detection reagent 1 125mL; ECL detection reagent 2 125mL |
| Product Type | Labeling and Detection System |
| Quantity | Labels 5 μg Nucleic Acid |
| Test Time | 20 min. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.