Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ ECL™ Prime Western Blotting Detection Reagent
GSA_VA

Catalog No. 45010090 Shop All Cytiva Products
Change view
Click to view available options
Sufficient For:
1000cm2 membrane
3000cm2 membrane
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
45-010-090 3000cm2 membrane
45-002-401 1000cm2 membrane
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-010-090 Supplier Cytiva Supplier No. RPN2236
Get an Extra Life Sciences Product with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Solution A (luminol solution) and Solution B (peroxide solution)

Enables the use of highly diluted primary and secondary antibodies without reducing sensitivity. Cytiva Lifescience™ Amersham™ ECL™ Prime Western Blotting Detection Reagent produces high signal intensity and sensitivity.

p>
  • Stable signal enables multiple exposures and makes reagent suitable for large experimental series
  • Provides convenient handling time between detection and completion of experiment
  • Optimized for imaging with ImageQuant™ LAS 4000 (CCD-based imaging), Amersham Hybond™ P (PVDF) membranes and compatible with Amersham Hyperfilm™, Amersham Hybond- ECL (nitrocellulose) membranes
  • Also compatible with Rainbow™ Molecular Weight Markers and Amersham ECL DualVue™ Western Blotting Markers

Education and research

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type ECL Western Blot Detection Reagent
For Use With (Application) Sensitive Quantitative or Confirmatory Western Blot
Sensitivity ∼1 pg
Sufficient For 3000cm2 membrane
Content And Storage Solution A (Luminol solution, 3 x 50mL) and Solution B (Peroxide solution, 3 x 50mL), 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.