Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ ECL™ Western Blotting Detection Reagents
GSA_VA

Catalog No. 45000878 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
62.5 mL
125 mL
250 mL
375 mL
Sufficient For:
1000 cm2 Membrane
2000 cm2 Membrane
4000 cm2 Membrane
6000 cm2 Membrane
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
45-000-878 62.5 mL 1000 cm2 Membrane
45-001-160 375 mL 6000 cm2 Membrane
45-000-885 125 mL 2000 cm2 Membrane
45-000-875 250 mL 4000 cm2 Membrane
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 45-000-878 Supplier Cytiva Supplier No. RPN2109
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use for all routine Western blotting. Cytiva Life Sciences™ Amersham™ ECL™ Western Blotting Detection Reagents are the world's first commercially available chemiluminescent detection reagents and still one of the most widely used.

  • Detectable on Hyperfilm™ ECL or with CCD imager
  • Uses enhanced luminol-based detection which is suitable for all routine Western Blotting

Western Blotting

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type ECL Western Blot Detection Reagent
For Use With (Application) Western Blot
Quantity 62.5 mL
Sensitivity ∼10 pg
Sufficient For 1000 cm2 Membrane
Content And Storage 62.5mL detection reagent, 162.5mL detection reagent 2. 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.