Learn More
Cytiva Amersham™ ECL™ Western Blotting System
GSA_VA
Catalog No. 45000877 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
Ea.
Catalog No. Quantity
45-000-877 Ea.
1 options
Catalog No. 45-000-877 Supplier Cytiva Supplier No. RPN2108
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Includes
Anti-mouse HRP conjugate, 100μL; anti-rabbit HRP conjugate, 100μL; ECL Detection Reagents 1 and 2, 62.5mL each; Blocking agent, 5g
Detect mouse and rabbit primary antibodies on Western blots. Cytiva Life Sciences™ Amersham™ ECL™ Western Blotting System uses HRP-conjugated secondary antibodies and ECL reagents for detection.
  • Provides rapid, sensitive results using simple protocols
  • Designed for use with nitrocellulose and PVDF membranes
  • Up to 10 times more sensitive than colorimetric methods
  • Results obtained on X-ray film provide hard copy data and avoid fading associated with colorimetric detection methods
  • Membranes can be reprobed up to 10 times
  • Linear response in the range 0.2 to 2 O.D. units using pre-flashed Hyperfilm™ ECL
  • Non-radioactive
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Western Blot
Quantity Ea.
Content And Storage 100μL Anti-Mouse HRP conjugate, 100μL Anti-Rabbit HRP conjugate, 62.5mL ECL™ detection reagent 1, 62.5mL ECL™ detection reagent 2, 25g blocking agent, sufficient for 10 blots (10 x 10)