Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva™ Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Rolls
GSA_VA

Catalog No. 45004003 Shop All Cytiva Products
Change view
Click to view available options
Width (Metric):
150 mm
20 cm
30 cm
Pore Size:
0.2 μm
0.45 μm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric) Pore Size
45-004-003 30 cm 0.45 μm
45-004-004 30 cm 0.2 μm
45-004-008 20 cm 0.45 μm
45-004-009 20 cm 0.2 μm
45-004-013 150 mm 0.45 μm
45-004-014 150 mm 0.2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 45-004-003 Supplier Cytiva™ Supplier No. 10600003
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use for the best results with fluorescence Western blotting detection applications. Cytiva Life Sciences™ Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.

Supplied in rolls.

  • Low background enables a dynamic range for complicated samples
  • Suitable for Western blots, dot-blot assays, and other protein or nucleic acid methods
  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 4 m
Width (Metric) 30 cm
For Use With (Application) Western blots, dot-blot assays, and other protein or nucleic acid methods
Format Roll
Material 100% Pure Nitrocellulose
Pore Size 0.45 μm
Dimensions (LxW) 30 cm x 4 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.