Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Rolls

Catalog No. 45004014 Shop All Cytiva Products
Change view
Click to view available options
Width (Metric):
150 mm
20 cm
30 cm
Pore Size:
0.2 μm
0.45 μm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric) Pore Size
45-004-014 150 mm 0.2 μm
45-004-004 30 cm 0.2 μm
45-004-003 30 cm 0.45 μm
45-004-008 20 cm 0.45 μm
45-004-009 20 cm 0.2 μm
45-004-013 150 mm 0.45 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 45-004-014 Supplier Cytiva Supplier No. 10600014
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use for the best results with fluorescence Western blotting detection applications. Cytiva Life Sciences™ Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.

Supplied in rolls.

  • Low background enables a dynamic range for complicated samples
  • Suitable for Western blots, dot-blot assays, and other protein or nucleic acid methods
  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 4 m
Width (Metric) 150 mm
For Use With (Application) Western blots, dot-blot assays, and other protein or nucleic acid methods
Format Roll
Material 100% Pure Nitrocellulose
Pore Size 0.2 μm
Dimensions (LxW) 150 mm x 4 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.