Learn More
Cytiva Amersham™ Streptavidin-Fluor Conjugates
Catalog No. 45000731 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
Cy3-Streptavidin
Cy5-Streptavidin
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
45-000-731 Cy3-Streptavidin
45-000-734 Cy5-Streptavidin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-731 Supplier Cytiva Supplier No. PA43001
Add to Cart
Add to Cart
Use for in situ hybridization and flow cytometry. Cytiva Life Sciences™ Amersham™ Streptavidin-Fluor Conjugates consist of streptavidin conjugated to a range of fluorescent dyes.
  • High specificity reagents
Applications:
in situ hybridization, flow cytometry
TRUSTED_SUSTAINABILITY
Specifications
Formulation This product is supplied as a lyophilized solid in Phosphate-buffered Saline (0.02M Potassium Phosphate, 0.15M NaCl) pH 7.2, containing 1%(w/v) Bovine Serum Albumin and 0.1% (w/v) Sodium Azide, reconstitute with 1mL of deionized Water, dispense into suitable aliquots and freeze, avoid repeated freeze-thaw cycles
For Use With (Application) Suitable for use in the Fluorescent analysis of Biotinylated samples
Quantity 1 mg
Product Type Cy3-Streptavidin
Kit Contents Lyophilized solid in phosphate-buffered saline (0.02M potassium phosphate, 0.15M NaCl) pH 7.2, containing 1%(w/v) bovine serum albumin and 0.1% (w/v) sodium azide, reconstitute with 1mL of deionized water, dispense into suitable aliquots and freeze, avoid repeated freeze-thaw cycles