Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMFR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111746
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111746 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111746 Supplier Invitrogen™ Supplier No. PA5111746
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSPVERPSSD QEEGETSAQT ERVPLDLSPR LEETLDFGEV EVEPSEVEDF EARGSRFSKS ADERQRMLVQ RKDELLQQAR KRFLNKS Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 90%.

Autocrine motility factor is a tumor motility-stimulating protein secreted by tumor cells. The protein encoded by this gene is a glycosylated transmembrane protein and a receptor for autocrine motility factor. The receptor, which shows some sequence similarity to tumor protein p53, is localized to the leading and trailing edges of carcinoma cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMFR
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AMFR
Gene Accession No. Q9UKV5
Gene Alias AMF receptor; Amfr; Autocrine motility factor receptor; autocrine motility factor receptor, E3 ubiquitin protein ligase; E3 ubiquitin-protein ligase AMFR; gp78; RING finger protein 45; RING-type E3 ubiquitin transferase AMFR; RNF45
Gene Symbols AMFR
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human AMFR. Recombinant protein control fragment (Product #RP-108126).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 267
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.