Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (±)-AMG 487
GREENER_CHOICE

Catalog No. 448710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-871-0 10 mg
44-875-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-871-0 Supplier Tocris Bioscience™ Supplier No. 4487/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

CXCR3 antagonist; inhibits cell migration and metastasis

(±)-AMG 487 is an antagonist of CXCR3; inhibits binding of 125I-IP-10 and 125I-ITAC to CXCR3 (IC50 values are 8.0 and 8.2 nM respectively). Inhibits CXCR3-mediated cell migration by the chemokines IP-10, ITAC and MiG in vitro (IC50 values are 8, 15 and 36 nM respectively). Also shown to inhibit lung metastasis in a mouse model of metastatic breast cancer.

This product is racemic.

Chemical Identifiers

CAS 947536-03-0
Molecular Formula C32H28F3N5O4

Specifications

Quantity 10 mg
Formula Weight 603.59
Purity >98%
Molecular Weight (g/mol) 603.602
Chemical Name or Material (±)-AMG 487
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.