Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AMG 510
GREENER_CHOICESDP

Catalog No. 771350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-135-0 50 mg
77-131-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 77-135-0 Supplier Tocris Bioscience™ Supplier No. 7713/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective KRASG12C inhibitor

AMG 510 is a potent and selective covalent KRASG12C inhibitor (IC50 = 90 nM). Also a potent inhibitor of phospho-ERK (IC50 = 68 nM). In an in vivo model of non-small cell lung cancer, AMG 510 causes regression of tumors, an increase in T-cell recruitment and a pro-inflammatory tumor microenvironment. Synergises with MEK inhibitors and Carboplatin (Cat. No. 2626) and is orally bioavailable.

Specifications

Chemical Name or Material (1 R)-6-Fluoro-7-(2-fluoro-6-hydroxyphenyl)-1-[4-methyl-2-(1-methylethyl)-3-pyridinyl]-4-[(2 S)-2-methyl-4-(1-oxo-2-propen-1-yl)-1-piperazinyl]pyrido[2,3-d]pyrimidin-2(1 H)-one
CAS 2252403-56-6
Quantity 50 mg
Target Ras GTPase Inhibitors
Molecular Formula C30H30F2N6O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.