Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AMG 517
GREENER_CHOICESDP

Catalog No. 599550 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-955-0 50 mg
59-951-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 59-955-0 Supplier R&D Systems™ Supplier No. 5995/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent TRPV1 antagonist

Potent TRPV1 antagonist (IC50 = 0.9 nM). Inhibits capsaican-induced flinching and reverses analgesia in an inflammatory pain model in rats. Orally bioavailable.

Chemical Identifiers

CAS 659730-32-2
Molecular Formula C20H13F3N4O2S

Specifications

Quantity 50 mg
Solubility Soluble to 100mM in DMSO
Product Line Tocris
Formula Weight 430.41
Purity >98%
Molecular Weight (g/mol) 430.405
Chemical Name or Material N-[4-[[6-[4-(Trifluoromethyl)phenyl]-4-pyrimidinyl]oxy]-2-benzothiazolyl]acetamide
Recommended Storage Store at 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.