Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AMG 9810
GREENER_CHOICE

Catalog No. 231610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-161-0 10 mg
23-165-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-161-0 Supplier Tocris Bioscience™ Supplier No. 2316/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective, competitive antagonist of TRPV1

Potent and selective, competitive vanilloid TRPV1 receptor antagonist (IC50 = 17 nM). Inhibits capsaicin-, proton-, heat- and endogenous ligand-induced activation of human and rat recombinant TRPV1 receptors. Displays antihyperalgesic properties in a rat model of inflammatory pain.

Chemical Identifiers

CAS 545395-94-6
Molecular Formula C21H23NO3

Specifications

Quantity 10 mg
Formula Weight 337.42
Purity >98%
Molecular Weight (g/mol) 337.419
Chemical Name or Material AMG 9810
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.