Learn More
EVOS™ EVOS™ Onstage Vessel Holder, Multi-Well Plates
Catalog No. 12563551
Change view
Click to view available options
Quantity:
1 each
Catalog No. Quantity
12-563-551 1 each
1 options
Catalog No. 12-563-551 Supplier EVOS™ Supplier No. AMEPVH028
Add to Cart
Add to Cart
For use with EVOS FL Auto Digital Inverted Fluorescence System (Cat. No. 12-463-377).
Onstage Vessel Holders
  • Accommodate a variety of flasks, plates and dishes
  • Maximize your choice of cell culture vessels when conducting time-lapse imaging experiments using the EVOS FL Auto Imaging System equipped with onstage incubator
  • Allow a perfect fit of your microwell plates or cell culture flasks/dishes to the master plate of the onstage incubator for increased precision in sample alignment
  • With a snug and secure fit, yet readily switched, these vessel holders make time-lapse imaging convenient and easy
TRUSTED_SUSTAINABILITY
Specifications
Type Vessel Holder
Product Line EVOS
For Use With (Equipment) EVOS™ FL Auto Imaging System
Holds Holds one multi-well plate
Quantity 1 each
How should the EVOS vessel holders be stored?
Store the EVOS vessel holders flat and protected from any contact with heavy objects as they can be distorted. Once the holders are distorted and no longer lay flat on the stage, they cannot be used. Store them individually in their own lidded plastic containers or small sturdy boxes.
How should I clean my EVOS vessel holders?
Wipe the vessel holder with paper towels or Kimwipes tissues dampened with 70% ethanol or 4,000 ppm hydrogen peroxide (H2O2).
I have an EVOS vessel that does not fit into any of the vessel holders that you offer. What are my options?
We do offer an option for making custom vessel holders. Please contact your Sales Representative for initiating a custom order.
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.