Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMH Monoclonal Antibody (14), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543047
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543047 100 μL
PIMA543483 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543047 Supplier Invitrogen™ Supplier No. MA543047
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Human AMH

Anti mullerian hormone (AMH) is a member of the TGF beta superfamily. It is secreted as a homodimeric 140 kDa disulfide linked precursor that is cleaved to release the mature 30 kDa homodimer. Originally classified as a fetal testicular hormone that inhibits Mullerian duct development, AMH is expressed post natally by immature Sertoli cells, and to a lesser degree by granulosa cells. AMH plays a role in testicular differentiation and in the regulation of ovarian follicle growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMH
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 14
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene AMH
Gene Accession No. P03971
Gene Alias AMH; Anti - Mullerian hormone (Mulerian inhibiting substance); anti-Muellerian hormone; anti-Mullerian hormone; MIF; MIS; Muellerian-inhibiting factor; muellerian-inhibiting substance; Mullerian inhibiting factor; Mullerian inhibiting substance
Gene Symbols AMH
Host Species Mouse
Immunogen Recombinant Human AMH Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.