Learn More
Invitrogen™ AMH Monoclonal Antibody (4-1#)

Description
AMH Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | AMH |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 4-1# |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | AMH |
| Gene Accession No. | P03971, P03972 |
| Gene Alias | AMH; Anti - Mullerian hormone (Mulerian inhibiting substance); anti-Muellerian hormone; anti-Mullerian hormone; MIF; MIS; Muellerian-inhibiting factor; muellerian-inhibiting substance; Mullerian inhibiting factor; Mullerian inhibiting substance |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.