Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMH Recombinant Rabbit Monoclonal Antibody (003), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

$293.50 - $565.00

Specifications

Antigen AMH
Clone 003
Concentration 1 mg/mL
Applications ELISA
Classification Recombinant Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIMA543369
View Documents
Invitrogen™
MA543369
50 μL
Each for $293.50
Only null left
Add to Cart
 
PIMA543056
View Documents
Invitrogen™
MA543056
100 μL
Each for $565.00
Only null left
Add to Cart
 
Description

Description

Specificity: Human AMH

Anti mullerian hormone (AMH) is a member of the TGF beta superfamily. It is secreted as a homodimeric 140 kDa disulfide linked precursor that is cleaved to release the mature 30 kDa homodimer. Originally classified as a fetal testicular hormone that inhibits Mullerian duct development, AMH is expressed post natally by immature Sertoli cells, and to a lesser degree by granulosa cells. AMH plays a role in testicular differentiation and in the regulation of ovarian follicle growth.
Specifications

Specifications

AMH
1 mg/mL
Recombinant Monoclonal
Liquid
RUO
PBS with no preservative
AMH; Anti - Mullerian hormone (Mulerian inhibiting substance); anti-Muellerian hormone; anti-Mullerian hormone; MIF; MIS; Muellerian-inhibiting factor; muellerian-inhibiting substance; Mullerian inhibiting factor; Mullerian inhibiting substance
AMH
IgG
Protein A
Antibody
003
ELISA
Unconjugated
Rabbit
Human
P03971
268
Recombinant Human AMH Protein.
Primary
-20°C, Avoid Freeze/Thaw Cycles
AMH
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.