Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMICA/JAML Antibody (401901) [Biotin], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB34491B
Description
AMICA/JAML Monoclonal antibody specifically detects AMICA/JAML in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
AMICA/JAML | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
120425 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, CyTOF | |
401901 | |
Flow Cytometry, CyTOF-ready | |
adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638 | |
Mouse myeloma cell line NS0-derived recombinant human AMICA/JAML, Leu20-Leu275, Accession # Q86YT9 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction