Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMICA/JAML Antibody (401901), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB34491MFV610
Description
AMICA/JAML Monoclonal antibody specifically detects AMICA/JAML in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| AMICA/JAML | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 120425 | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 401901 | |
| Flow Cytometry, CyTOF-ready | |
| adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638 | |
| Mouse myeloma cell line NS0-derived recombinant human AMICA/JAML, Leu20-Leu275, Accession # Q86YT9 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction