Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMICA/JAML Antibody (4E10), Alexa Fluor™ 350, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP143309AF350
Description
AMICA/JAML Monoclonal specifically detects AMICA/JAML in Mouse samples. It is validated for Flow Cytometry.Specifications
AMICA/JAML | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
JAML | |
Synthetic peptide corresponding to residues C(267) D G I L G D N F R P T Q(279) of mouse CARP. | |
0.1 mL | |
120425.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
40000000000 | |
Flow Cytometry | |
adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638 | |
Armenian Hamster | |
Protein A or G purified | |
Primary | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction