Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMICA/JAML Antibody (4E10), DyLight 680, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP143309FR
Description
AMICA/JAML Monoclonal specifically detects AMICA/JAML in Mouse samples. It is validated for Flow Cytometry.Specifications
AMICA/JAML | |
Monoclonal | |
DyLight 680 | |
adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638 | |
Armenian Hamster | |
Affinity Purified | |
RUO | |
120425 | |
Store at 4C in the dark. |
Flow Cytometry | |
40000000000 | |
Flow Cytometry | |
AMICA1 | |
The immunogen for this antibody was JAML. | |
0.1 mL | |
Primary | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction