Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMICA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100845 Supplier Invitrogen™ Supplier No. PA5100845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total JAML1.

AMICA transmembrane protein of the plasma membrane of leukocytes that control their migration and activation through interaction with CXADR, a plasma membrane receptor found on adjacent epithelial and endothelial cells. The interaction between both receptors mediates the activation of gamma-delta T-cells, a subpopulation of T-cells residing in epithelia and involved in tissue homeostasis and repair. Upon epithelial CXADR-binding, JAML induces downstream cell signaling events in gamma-delta T-cells through PI3-kinase and MAP kinases. It results in proliferation and production of cytokines and growth factors by T-cells that in turn stimulate epithelial tissues repair. It also controls the transmigration of leukocytes within epithelial and endothelial tissues through adhesive interactions with epithelial and endothelial CXADR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMICA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene JAML
Gene Accession No. Q86YT9
Gene Alias adhesion molecule AMICA; adhesion molecule interacting with CXADR antigen 1; adhesion molecule, interacts with CXADR antigen 1; AMICA; Amica1; Crea7; CREA7-1; CREA7-4; Dendritic cell-specific protein CREA7; Dendritic cell-specific protein CREA7-1; dendritic-cell specific protein Crea7; dendritic-cell specific protein CREA7-1; dendritic-cell specific protein CREA7-4; Gm638; Jaml; junction adhesion molecule like; junctional adhesion molecule-like; mCrea7; UNQ722/PRO1387
Gene Symbols JAML
Host Species Rabbit
Immunogen A synthesized peptide derived from human JAML(Accession Q86YT9), corresponding to amino acid residues E349-F394.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120425, 315610
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.