Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Amicon™ Pro Purification System with Ultra-0.5 Device, 50 Kda

Catalog No. p-7232202 Shop All MilliporeSigma Products
Click to view available options
Quantity:
24Pack
Includes:
24 Amicon Ultra NMWCO 50 KDa filters, 24 Exchange devices, and 24 Collection tubes with cap

Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.

Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.

The Amicon™ Pro Purification System includes: Amicon Ultra NMWCO filters, Exchange devices, and Collection tubes with cap.
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
MWCO 50 kDa
Sterility Non-sterile
Housing Material Regenerated Cellulose (RC)
Includes 24 Amicon Ultra NMWCO 50 KDa filters, 24 Exchange devices, and 24 Collection tubes with cap
Quantity 24Pack
Type Centrifugal Filters
Volume (Metric) 10 mL
Length (Metric) 12 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.