Learn More
Invitrogen™ AMID Monoclonal Antibody (3A11)

Description
Peptide Sequence: VRQEVKEILL RKGVQLLLSE RVSNLEELPL NEYREYIKVQ TDKGTEVATN LVILCTGIKI NSSAYRKAFE SRLASSGALR VNEHLQVEGH SNVYAIGDCA.
Specifications
Specifications
| Antigen | AMID |
| Applications | ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 3A11 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | AIFM2 |
| Gene Accession No. | Q9BRQ8 |
| Gene Alias | 5430437E11Rik; AIFM2; Amid; apoptosis inducing factor, mitochondria associated 2; apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer of death; apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death; apoptosis-inducing factor 2; Apoptosis-inducing factor homologous mitochondrion-associated inducer of death; apoptosis-inducing factor, mitochondrion-associated 2; apoptosis-inducing factor, mitochondrion-associated, 2; apoptosis-inducing factor-like mitchondrion-associated inducer of death; apoptosis-inducing factor-like mitochondrion-associated inducer of death; D730001I10Rik; MBP2; MBPH; MGC126662; MGC141971; p53-responsive gene 3 protein; PRG3; RP11-367H5.2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.