Learn More
Invitrogen™ AMID Polyclonal Antibody

Description
Immunogen sequence: NLVILCTGIK INSSAYRKAF ESRLASSGAL RVNEHLQVEG HSNVYAIGDC ADVRTPKMAY LAGLHANIAV ANIVNSVKQR PLQAYKPGAL TFLLSMGRND GVGQISGFYV GRLMVRLTKS RDLFVSTSWK TM Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 86%.
Specifications
Specifications
| Antigen | AMID |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | AIFM2 |
| Gene Accession No. | Q9BRQ8 |
| Gene Alias | 5430437E11Rik; AIFM2; Amid; apoptosis inducing factor, mitochondria associated 2; apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer of death; apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death; apoptosis-inducing factor 2; Apoptosis-inducing factor homologous mitochondrion-associated inducer of death; apoptosis-inducing factor, mitochondrion-associated 2; apoptosis-inducing factor, mitochondrion-associated, 2; apoptosis-inducing factor-like mitchondrion-associated inducer of death; apoptosis-inducing factor-like mitochondrion-associated inducer of death; D730001I10Rik; MBP2; MBPH; MGC126662; MGC141971; p53-responsive gene 3 protein; PRG3; RP11-367H5.2 |
| Gene Symbols | AIFM2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.