Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amido Black 10B

Catalog No. p-7076511
Click to view available options
Quantity:
25 g
100 g

Used as a histological, protein, and nucleic acid dye used in electrophoresis. | CAS: 1064-48-8 | C22H14N6Na2O9S2 | 616.487 g/mol

Amido Black 10B is used as a histological, protein, and nucleic acid dye used in electrophoresis. Naphthol Blue Black is an important and commonly used industrial dye. This dye is also used in rapid staining of protein bands on nitrocellulose membranes such as Western blots. Naphthol Blue Black can be utilized as a nucleic acid stain in SDS-Page gels. Additionally this compound has been untilized in cell proliferation assays and histochemical studies. It is also used in criminal investigations to detect blood present with latent fingerprints. They are also used in paints, inks, plastics and leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1064-48-8
Molecular Formula C22H14N6Na2O9S2
Molecular Weight (g/mol) 616.487
MDL Number MFCD00004017
InChI Key HKBVRFLHNUEVRO-UHFFFAOYSA-L
Synonym C.I. 20470,Naphthalene Black 10B
PubChem CID 44134531
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Beilstein 5374263
Solubility Information It is soluble in water (10mg/ml at 25°C),and ethanol.
Formula Weight 616.49
Packaging Glass bottle
Sensitivity Hygroscopic
Quantity 25 g
Physical Form Powder
Chemical Name or Material Amido Black 10B
Hazard Category H315-H317-H319-H335-H372-H500
Precautionary Statement P261-P264b-P270-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P314-P332+P313-P362-P501c
EINECSNumber 213-903-1
RTECSNumber QJ6196000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.