Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amidosulfonic acid, 99.99% (metals basis)

Catalog No. AA4451036
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 5329-14-6 | H3NO3S | 97.088 g/mol

Amidosulfonic acid is mainly a precursor to sweet-tasting compounds. Reaction with cyclohexylamine followed by addition of NaOH gives C6H11NHSO3Na, sodium cyclamate. Sulfamic acid is used as an acidic cleaning agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
Molecular Weight (g/mol) 97.088
MDL Number MFCD00011603
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
Synonym amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
PubChem CID 5987
ChEBI CHEBI:9330
IUPAC Name sulfamic acid
SMILES NS(=O)(=O)O

Specifications

pH 1.2
Melting Point ∼205°C (decomposition)
Density 2.12 g/mL
Odor Odorless
Quantity 500 g
Physical Form Crystalline
UN Number UN2967
Merck Index 148921
Solubility Information Soluble in water.
Formula Weight 97.09
Percent Purity 99.99%
Assay Metals basis
Chemical Name or Material Amidosulfonic acid
Show More Show Less
Hazard Category H290-H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P234-P264b-P280g-P280i-P302+P352-P305+P351+P338-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: SULFAMIC ACID
EINECSNumber 226-218-8
RTECSNumber W05950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.