Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Amifostine trihydrate

Catalog No. AC463102500
Click to view available options
Quantity:
250 mg
1 g

CAS: 112901-68-5 | C5H21N2O6PS | 268.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112901-68-5
Molecular Formula C5H21N2O6PS
Molecular Weight (g/mol) 268.26
InChI Key TXQPXJKRNHJWAX-UHFFFAOYSA-N
IUPAC Name ({2-[(3-aminopropyl)amino]ethyl}sulfanyl)phosphonic acid trihydrate
SMILES O.O.O.NCCCNCCSP(O)(O)=O

Specifications

Quantity 250 mg
Formula Weight 268.27
Chemical Name or Material Amifostine trihydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.