Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMIGO Antibody (S86-36), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222413G
Description
AMIGO Monoclonal specifically detects AMIGO in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
AMIGO | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
AMIGO1 | |
Fusion protein amino acids 554-574 (cytoplasmic C-terminus) of human AMIGO-1 | |
0.1 mL | |
57463 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S86-36 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
adhesion molecule with Ig-like domain 1, ALI2, Alivin-2, AMIGO-1, AMIGOamphoterin-induced protein 1, amphoterin-induced gene and ORF, KIAA1163 | |
Mouse | |
Protein G purified | |
Primary | |
Detects approx 60-80kDa depending on maturity/glycosylation. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction