Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMIGO1 Monoclonal Antibody (L86/33), FITC
GREENER_CHOICE

Catalog No. PIMA545620
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545620 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA545620 Supplier Invitrogen™ Supplier No. MA545620
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

1 μg/mL of MA5-45620 was sufficient for detection of AMIGO-1 in 20 μg of rat brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 60-80kDa depending on maturity/glycosylation. This antibody was formerly sold as clone S86-36.

Promotes growth and fasciculation of neurites from cultured hippocampal neurons. May be involved in fasciculation as well as myelination of developing neural axons. May have a role in regeneration as well as neural plasticity in the adult nervous system. May mediate homophilic as well as heterophilic cell-cell interaction and contribute to signal transduction through its intracellular domain. Assembled with KCNB1 modulates the gating characteristics of the delayed rectifier voltage-dependent potassium channel KCNB1. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMIGO1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone L86/33
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene Amigo1
Gene Accession No. Q80ZD7, Q80ZD8, Q86WK6
Gene Alias adhesion molecule with Ig like domain 1; adhesion molecule with Ig-like domain 1; ali2; alivin 2; alivin-2; AMIGO; AMIGO 1; AMIGO1; AMIGO-1; amigo1 {ECO:0000250; amphoretin-induced gene and ORF; amphoterin induced gene and ORF (Amigo); amphoterin-induced gene and ORF; amphoterin-induced protein 1; KIAA1163; MGC25558; OTTHUMP00000013379; RP23 89M15.6; transmembrane protein AMIGO; UniProtKB:Q80ZD8}
Gene Symbols Amigo1
Host Species Mouse
Immunogen Fusion proteinamino acids 394-492 (cytoplasmic C-terminus) of mouse AMIGO-1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229715, 295365, 57463
Target Species Human, Mouse, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.