Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMIGO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561164
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561164 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561164 Supplier Invitrogen™ Supplier No. PA561164
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSVMDFQEDL YCMNSKKLHN VFNLSFLNCG EYKERAWEAH LGDTLIIKCD TKQQGMTKVW VTPSNERVLD EVTNGTVSVS KDG Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 81%.

Promotes growth and fasciculation of neurites from cultured hippocampal neurons. May be involved in fasciculation as well as myelination of developing neural axons. May have a role in regeneration as well as neural plasticity in the adult nervous system. May mediate homophilic as well as heterophilic cell-cell interaction and contribute to signal transduction through its intracellular domain. Assembled with KCNB1 modulates the gating characteristics of the delayed rectifier voltage-dependent potassium channel KCNB1. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMIGO1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Amigo1
Gene Accession No. Q86WK6
Gene Alias adhesion molecule with Ig like domain 1; adhesion molecule with Ig-like domain 1; ali2; alivin 2; alivin-2; AMIGO; AMIGO 1; AMIGO1; AMIGO-1; amigo1 {ECO:0000250; amphoretin-induced gene and ORF; amphoterin induced gene and ORF (Amigo); amphoterin-induced gene and ORF; amphoterin-induced protein 1; KIAA1163; MGC25558; OTTHUMP00000013379; RP23 89M15.6; transmembrane protein AMIGO; UniProtKB:Q80ZD8}
Gene Symbols Amigo1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AMIGO1. Recombinant protein control fragment (Product #RP-95716).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57463
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.